missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzoic Acid, TRC

Product Code. 30387790
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387790 50 g -
30381195 100 g 100g
30378792 25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30387790 Supplier Toronto Research Chemicals Supplier No. TRCD47091550G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-84-1
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
Synonym 4-(N,N-Dimethylamino)benzoic Acid, N,N-Dimethyl-4-aminobenzoic Acid, N,N-Dimethyl-p-aminobenzoic Acid, NSC 16596, p-(Dimethylamino)benzoic Acid
IUPAC Name 4-(dimethylamino)benzoic acid
SMILES CN(C)c1ccc(cc1)C(=O)O

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H11NO2/c1-10(2)8-5-3-7(4-6-8)9(11)12/h3-6H,1-2H3,(H,11,12)
Formula Weight 165.079
Chemical Name or Material 4-(Dimethylamino)benzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.