missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)benzaldehyde, TRC

Product Code. 30133542
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133542 50 g 50g
30004438 100 g 100g
30010062 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133542 Supplier Toronto Research Chemicals Supplier No. TRCD46126050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9 H11 N O
Molecular Weight (g/mol) 149.19
Synonym Benzaldehyde, 4-(dimethylamino)-, Benzaldehyde, p-(dimethylamino)- (8CI), 4-(Dimethylamino)benzaldehyde, 4-(Dimethylamino)benzenecarbonal, 4-(N,N-Dimethylamino)benzaldehyde, 4-Formyl-N,N-dimethylaniline, N,N-Dimethyl-4-aminobenzaldehyde, N,N-Dimethyl-4-formylaniline, N,N-Dimethyl-p-aminobenzaldehyde, NSC 5517, Named reagents and solutions, Ehrlich's, p-(Dimethylamino)benzaldehyde, p-(N,N-Dimethylamino)benzaldehyde, p-DAB, p-Formyl-N,N-dimethylaniline, p-Formyldimethylaniline
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)c1ccc(C=O)cc1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3
Formula Weight 149.0841
Chemical Name or Material 4-(Dimethylamino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.