missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)antipyrine, 98+%

Product Code. 11469417
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469417 50 g 50g
11479417 250 g 250g
11459417 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469417 Supplier Thermo Scientific Alfa Aesar Supplier No. B21436.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58-15-1 | C13H17N3O | 231.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-15-1
Molecular Formula C13H17N3O
Molecular Weight (g/mol) 231.30
MDL Number MFCD00003142
InChI Key RMMXTBMQSGEXHJ-UHFFFAOYSA-N
Synonym aminophenazone, amidopyrine, aminopyrine, 4-dimethylaminoantipyrine, dipyrine, amidazophen, amidophen, aminopyrin, pyramidon, amidophenazone
PubChem CID 6009
ChEBI CHEBI:160246
IUPAC Name 4-(dimethylamino)-1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CN(C)C1=C(C)N(C)N(C1=O)C1=CC=CC=C1

Specifications

Melting Point 105°C to 109°C
Odor Odorless
Quantity 50 g
UN Number UN2811
Merck Index 14,474
Formula Weight 231.3
Percent Purity ≥98%
Chemical Name or Material 4-(Dimethylamino)antipyrine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.