missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethylamino)-3-hydroxybutanoic Acid, TRC

Product Code. 30381714
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381714 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381714 Supplier Toronto Research Chemicals Supplier No. TRCD462025100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Dimethylamino)-3-hydroxybutanoic Acid
CAS 542-06-3
Molecular Formula C6 H13 N O3
Quantity 100 mg
Synonym DL-Dimethylamino-3-hydroxybutyric Acid, DL-Norcarnitine, NSC 40237, Norcarnitine, (±)-4-(Dimethylamino)-3-hydroxybutyric Acid, (±)-4-(Dimethylamino)-3-hydroxybutanoic Acid
InChI Formula InChI=1S/C6H13NO3/c1-7(2)4-5(8)3-6(9)10/h5,8H,3-4H2,1-2H3,(H,9,10)
SMILES CN(C)CC(O)CC(=O)O
IUPAC Name 4-(dimethylamino)-3-hydroxybutanoic acid
Molecular Weight (g/mol) 147.172
Formula Weight 147.09
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.