missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Dimethylamino)-1,1-dimethoxybut-3-en-2-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | (CH3)2NCH=CHC(O)CH(OCH3)2 |
Quantity | 1 g |
Formula Weight | 173.21 |
Percent Purity | 97% |
Chemical Name or Material | 4-(Dimethylamino)-1, 1-dimethoxybut-3-en-2-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'