missing translation for 'onlineSavingsMsg'
Learn More

4-(Dimethoxymethyl)-2-methylpyrimidine, 97%, Thermo Scientific™

Product Code. 10168754
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10168754 25 g 25g
10089491 1 g 1g
10394912 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10168754 Supplier Thermo Scientific Maybridge Supplier No. SPB04845EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-33-5
Molecular Formula C8H12N2O2
Molecular Weight (g/mol) 168.20
MDL Number MFCD00085137
InChI Key VDXHJWMLYFVGDL-UHFFFAOYSA-N
PubChem CID 2799572
IUPAC Name 4-(dimethoxymethyl)-2-methylpyrimidine
SMILES COC(OC)C1=CC=NC(C)=N1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Synonym 4-dimethoxymethyl-2-methylpyrimidine, 2-methylpyrimidine-4-carboxaldehyde dimethylacetal, 4-dimethoxymethyl-2-methyl-pyrimidine, acmc-20a0b8, dimethoxy 2-methylpyrimidin-4-yl methane, pyrimidine,4-dimethoxymethyl-2-methyl, pyrimidine, 4-dimethoxymethyl-2-methyl
Formula Weight 168.19
Percent Purity 97%
Chemical Name or Material 4-(Dimethoxymethyl)-2-methylpyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.