missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)benzaldehyde, 97%, Maybridge

Product Code. 10425912
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10425912 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10425912 Supplier Thermo Scientific Maybridge Supplier No. TL00446DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 73960-07-3
CAS Min % 97.0
Density 1.302g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H6F2O2
Linear Formula F2CHOC6H4CHO
MDL Number MFCD00042252
Quantity 1 g
Synonym 4-difluoromethoxy benzaldehyde, 4-difluoromethoxy-benzaldehyde, p-difluoromethoxy benzaldehyde, p-difluoromethoxybenzaldehyde, benzaldehyde, 4-difluoromethoxy, 1-difluoromethoxy-4-formylbenzene, pubchem7137, acmc-1bb9r, akos bb62
InChI Key ZWCXOJYJJINQGU-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1C=O)OC(F)F
IUPAC Name 4-(difluoromethoxy)benzaldehyde
Molecular Weight (g/mol) 172.131
PubChem CID 2736986
Formula Weight 172.13
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.