missing translation for 'onlineSavingsMsg'
Learn More

4-(Difluoromethoxy)-2-fluorobenzyl bromide, 97%, Thermo Scientific™

Catalog Number 15447965
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1017779-33-7
C8H6BrF3O
255.03
MFCD09832360
JCIXDPWAAZBQKQ-UHFFFAOYSA-N
4-difluoromethoxy-2-fluorobenzyl bromide, 1-bromomethyl-4-difluoromethoxy-2-fluorobenzene, 4-bromomethyl-3-fluorophenoxy difluoromethane, 4-difluoromethoxy-2-fluorobenzyl bromide, jrd
46737676
1-(bromomethyl)-4-(difluoromethoxy)-2-fluorobenzene
FC(F)OC1=CC(F)=C(CBr)C=C1
This item is not returnable. View return policy
1017779-33-7
C8H6BrF3O
255.03
MFCD09832360
JCIXDPWAAZBQKQ-UHFFFAOYSA-N
4-difluoromethoxy-2-fluorobenzyl bromide, 1-bromomethyl-4-difluoromethoxy-2-fluorobenzene, 4-bromomethyl-3-fluorophenoxy difluoromethane, 4-difluoromethoxy-2-fluorobenzyl bromide, jrd
46737676
1-(bromomethyl)-4-(difluoromethoxy)-2-fluorobenzene
FC(F)OC1=CC(F)=C(CBr)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY
Refractive Index 1.505
Quantity 250 mg
UN Number UN3265
Solubility Information Not miscible or difficult to mix.
Formula Weight 255.03
Percent Purity 97%
Chemical Name or Material 4-(Difluoromethoxy)-2-fluorobenzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'