missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylcarbamoyl)benzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15459757
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15459757 5 g 5g
15449757 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459757 Supplier Thermo Scientific Alfa Aesar Supplier No. H52986.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 389621-80-1
Molecular Formula C11H16BNO3
Molecular Weight (g/mol) 221.06
MDL Number MFCD03411949
InChI Key ZCGVBHIMRVYWOH-UHFFFAOYSA-N
Synonym 4-n,n-diethylaminocarbonyl phenylboronic acid, 4-diethylcarbamoyl phenylboronic acid, 4-diethylcarbamoyl phenyl boronic acid, 4-diethylcarbamoyl benzeneboronic acid, boronic acid, 4-diethylamino carbonyl phenyl, 4-diethylaminocarbonyl phenylboronic acid, acmc-209j2q, amtb302, n,n-diethyl 4-boronobenzamide, 4-diethylcarbamoyl benzeneboronic acid
PubChem CID 2773375
IUPAC Name [4-(diethylcarbamoyl)phenyl]boronic acid
SMILES CCN(CC)C(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point 138°C to 142°C
Quantity 5 g
Formula Weight 221.06
Percent Purity 98%
Chemical Name or Material 4-(Diethylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.