missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminosalicylaldehyde, 99%

Product Code. 11342557
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342557 5 g 5g
11352557 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342557 Supplier Thermo Scientific Alfa Aesar Supplier No. L11659.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17754-90-4 | C11H15NO2 | 193.246 g/mol

4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminosalicylaldehyde is used in the synthesis of Schiff-base ligand by monocondensation with diaminomaleonitrile.

Solubility
Insoluble in water.

Notes
It is sensitive to air. Store at room temperature. Incompatible with strong bases and strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17754-90-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD00003326
InChI Key XFVZSRRZZNLWBW-UHFFFAOYSA-N
Synonym 4-diethylamino salicylaldehyde, 4-diethylamino-2-hydroxybenzaldehyde, 4-diethylaminosalicylaldehyde, 4-n,n-diethylamino salicylaldehyde, benzaldehyde, 4-diethylamino-2-hydroxy, p-diethylamino salicylaldehyde, unii-ou5nfu4681, 2-hydroxy-4-diethylaminobenzaldehyde, 4-n,n-diethylaminosalicylic aldehyde, 4-diethylamino-2-hydroxy-benzaldehyde
PubChem CID 87293
IUPAC Name 4-(diethylamino)-2-hydroxybenzaldehyde
SMILES CCN(CC)C1=CC(=C(C=C1)C=O)O

Specifications

Melting Point 60°C to 64°C
Quantity 5 g
Beilstein 2212234
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 193.25
Percent Purity 99%
Chemical Name or Material 4-Diethylaminosalicylaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.