missing translation for 'onlineSavingsMsg'
Learn More

4-Diethylaminobenzaldehyde, 97%

Product Code. 11419423
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419423 100 g 100g
11429423 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419423 Supplier Thermo Scientific Alfa Aesar Supplier No. A11825.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-21-8 | C11H15NO | 177.247 g/mol

4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Diethylaminobenzaldehyde is used in the preparation of 5-(4-diethylamino-benzylidene)-selenazolidine-2,4-dione by using aqueous methylamine as a reagent.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-21-8
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00003382
InChI Key MNFZZNNFORDXSV-UHFFFAOYSA-N
Synonym 4-diethylamino benzaldehyde, benzaldehyde, 4-diethylamino, p-diethylamino benzaldehyde, p-formyl-n,n-diethylaniline, 4-diethylamino-benzaldehyde, benzaldehyde, p-diethylamino, unii-s640xwl10h, p-n,n-diethylaminobenzaldehyde, p-diethylaminobenzaldehyde, 4-n,n-diethylamino benzaldehyde
PubChem CID 67114
ChEBI CHEBI:86194
IUPAC Name 4-(diethylamino)benzaldehyde
SMILES CCN(CC)C1=CC=C(C=C1)C=O

Specifications

Melting Point 39°C to 41°C
Boiling Point 173°C to 175°C (7 mmHg)
Flash Point 165°C (329°F)
Quantity 100 g
Beilstein 511102
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 177.25
Percent Purity 97%
Chemical Name or Material 4-Diethylaminobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.