missing translation for 'onlineSavingsMsg'
Learn More

4-(Diethylamino)butanenitrile, 97%, Thermo Scientific™

Product Code. 10358370 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10358370

Brand: Thermo Scientific Acros 235570010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5336-75-4
CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Density 0.8400g/mL
Boiling Point 89°C to 90°C (9.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C8H16N2
Linear Formula (CH3CH2)2NCH2CH2CH2CN
Refractive Index 1.4350 to 1.437
MDL Number MFCD00019893
Quantity 1 g
Specific Gravity 0.84
Synonym 4-diethylamino butanenitrile, 4-diethylaminobutyronitrile, gamma-diethylaminobutyronitrile, butanenitrile,4-diethylamino, butanenitrile, 4-diethylamino, 4-diethylamino butyronitrile, 4-diethylamino-butyronitrile
InChI Key NUVVGLXJJPTXRJ-UHFFFAOYSA-N
SMILES CCN(CC)CCCC#N
IUPAC Name 4-(diethylamino)butanenitrile
Molecular Weight (g/mol) 140.23
PubChem CID 79262
Formula Weight 140.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4-(Diethylamino)butanenitrile, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Diethylamino)butanenitrile
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.