missing translation for 'onlineSavingsMsg'
Learn More

4-(Dibutylamino)benzaldehyde, TRC

Product Code. 30380375
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380375 1 g 1g
30386494 500 mg -
30379168 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30380375 Supplier Toronto Research Chemicals Supplier No. TRCD4627731G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90134-10-4
Molecular Formula C15H23NO
Molecular Weight (g/mol) 233.35
Synonym 4-(Dibutylamino)benzaldehyde, 4-(Dibutylamino)benzyaldehyde, 4-(N,N-Dibutylamino)benzaldehyde, N,N-Dibutyl-4-aminobenzaldehyde, N,N-Dibutyl-4-formylaniline, p-(Dibutylamino)benzaldehyde, p-(N,N-Dibutylamino)benzaldehyde
SMILES CCCCN(CCCC)c1ccc(C=O)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H23NO/c1-3-5-11-16(12-6-4-2)15-9-7-14(13-17)8-10-15/h7-10,13H,3-6,11-12H2,1-2H3
Formula Weight 233.178
Chemical Name or Material 4-(Dibutylamino)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.