missing translation for 'onlineSavingsMsg'
Learn More

4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazineethanol, TRC

Product Code. 30136273
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136273 500 mg 500mg
30005647 100 mg 100mg
30008346 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136273 Supplier Toronto Research Chemicals Supplier No. TRCD418698500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 329216-67-3
Molecular Formula C19 H21 N3 O S
Molecular Weight (g/mol) 339.45
Synonym 1-Piperazineethanol, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, 4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazineethanol, 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol
IUPAC Name 2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethanol
SMILES OCCN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H21N3OS/c23-14-13-21-9-11-22(12-10-21)19-15-5-1-3-7-17(15)24-18-8-4-2-6-16(18)20-19/h1-8,23H,9-14H2
Formula Weight 339.1405
Percent Purity >95%
Chemical Name or Material 2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethanol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.