missing translation for 'onlineSavingsMsg'
Learn More

4-Desfluoro-2-Fluoro Droperidol, TRC

Product Code. 30009288
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
150 mg
Unit Size:
150mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009288 25 mg 25mg
30008281 150 mg 150mg
30006344 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009288 Supplier Toronto Research Chemicals Supplier No. TRCD29086525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1026015-45-1
Molecular Formula C22 H22 F N3 O2
Molecular Weight (g/mol) 379.4274
Synonym 2H-Benzimidazol-2-one, 1-[1-[4-(2-fluorophenyl)-4-oxobutyl]-1,2,3,6-tetrahydro-4-pyridinyl]-1,3-dihydro-, 1-[1-[4-(2-Fluorophenyl)-4-oxobutyl]-1,2,3,6-tetrahydro-4-pyridinyl]-1,3-dihydro-2H-Benzimidazol-2-one, 4-Desfluoro-2-Fluoro Droperidol
IUPAC Name 3-[1-[4-(2-fluorophenyl)-4-oxobutyl]-3,6-dihydro-2H-pyridin-4-yl]-1H-benzimidazol-2-one
SMILES Fc1ccccc1C(=O)CCCN2CCC(=CC2)N3C(=O)Nc4ccccc34

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H22FN3O2/c23-18-7-2-1-6-17(18)21(27)10-5-13-25-14-11-16(12-15-25)26-20-9-4-3-8-19(20)24-22(26)28/h1-4,6-9,11H,5,10,12-15H2,(H,24,28)
Formula Weight 379.1696
Chemical Name or Material 1-[1-[4-(2-Fluorophenyl)-4-oxobutyl]-1,2,3,6-tetrahydropyridin-4-yl]-1,3-dihydro-2H-benzimidazol-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.