missing translation for 'onlineSavingsMsg'
Learn More

4-Desethylamino 4-oxobrinzolamide, TRC

Product Code. 30387026
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30387026 25 mg
30381055 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30387026 Supplier Toronto Research Chemicals Supplier No. TRCD44967025MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154127-41-0
Molecular Formula C10H14N2O6S3
Molecular Weight (g/mol) 354.42
Synonym 3,4-Dihydro-2-(3-methoxypropyl)-4-oxo-2H-thieno[3,2-e]-1,2-thiazine-6-sulfonamide 1,1-Dioxide
SMILES COCCCN1CC(=O)c2cc(sc2S1(=O)=O)S(=O)(=O)N

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H14N2O6S3/c1-18-4-2-3-12-6-8(13)7-5-9(20(11,14)15)19-10(7)21(12,16)17/h5H,2-4,6H2,1H3,(H2,11,14,15)
Formula Weight 354.0014
Chemical Name or Material 4-Desethylamino 4-oxobrinzolamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.