missing translation for 'onlineSavingsMsg'
Learn More

4' '-Des-morpholino,-4' '-amino Momelotinib, TRC

Product Code. 30385093
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385093 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385093 Supplier Toronto Research Chemicals Supplier No. TRCD96570010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[2-(4-aminoanilino)pyrimidin-4-yl]-N-(cyanomethyl)benzamide
CAS 1841094-21-0
Molecular Formula C19 H16 N6 O
Quantity 10 mg
Synonym 4-[2-[(4-Aminophenyl)amino]-4-pyrimidinyl]-N-(cyanomethyl)benzamide
InChI Formula InChI=1S/C19H16N6O/c20-10-12-22-18(26)14-3-1-13(2-4-14)17-9-11-23-19(25-17)24-16-7-5-15(21)6-8-16/h1-9,11H,12,21H2,(H,22,26)(H,23,24,25)
SMILES Nc1ccc(Nc2nccc(n2)c3ccc(cc3)C(=O)NCC#N)cc1
IUPAC Name 4-[2-(4-aminoanilino)pyrimidin-4-yl]-N-(cyanomethyl)benzamide
Molecular Weight (g/mol) 344.37
Formula Weight 344.139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.