missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10199919 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10199919 1 g 1g
10704872 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10199919 Supplier Thermo Scientific Acros Supplier No. 438000010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 56354-57-5
Molecular Formula C12H15ClO2S
Molecular Weight (g/mol) 258.76
MDL Number MFCD00625713
InChI Key CREMYEDHKUWVTB-UHFFFAOYSA-N
Synonym 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
PubChem CID 4339442
IUPAC Name 4-cyclohexylbenzene-1-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 54.2°C to 55.7°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. as diethylsulfonamide (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.77
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Cyclohexylbenzenesulfonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Cyclohexylbenzenesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.