missing translation for 'onlineSavingsMsg'
Learn More

4-Cyclohexylbenzenesulfonyl chloride, 97%

Product Code. 15496457
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496457 1 g 1g
15406467 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496457 Supplier Thermo Scientific Alfa Aesar Supplier No. H33616.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56354-57-5 | C12H15ClO2S | 258.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56354-57-5
Molecular Formula C12H15ClO2S
Molecular Weight (g/mol) 258.76
MDL Number MFCD00625713
InChI Key CREMYEDHKUWVTB-UHFFFAOYSA-N
Synonym 4-cyclohexylbenzene-1-sulfonyl chloride, 4-cyclohexyl-benzenesulfonyl chloride, acmc-1awu9, chloro 4-cyclohexylphenyl sulfone, 4-cyclohexylphenylsulfonyl chloride, 4-cyclohexylbenzene-1-sulfonylchloride, 4-cyclohexyl-1-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyclohexyl
PubChem CID 4339442
IUPAC Name 4-cyclohexylbenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

Melting Point 52°C to 54°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 258.77
Percent Purity 97%
Chemical Name or Material 4-Cyclohexylbenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.