missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyclohexylacetophenone, 99%

Product Code. 11485776
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485776 100 g 100g
11465776 5 g 5g
11475776 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485776 Supplier Thermo Scientific Alfa Aesar Supplier No. B20838.22
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18594-05-3 | C14H18O | 202.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18594-05-3
Molecular Formula C14H18O
Molecular Weight (g/mol) 202.30
MDL Number MFCD00001453
InChI Key MSDQNIRGPBARGC-UHFFFAOYSA-N
Synonym 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone
PubChem CID 87715
SMILES CC(=O)C1=CC=C(C=C1)C1CCCCC1

Specifications

Melting Point 66°C to 70°C
Boiling Point 179°C to 181°C (14 mmHg)
Quantity 100 g
Beilstein 1962365
IUPAC Name 1-(4-cyclohexylphenyl)ethanone
Formula Weight 202.3
Percent Purity 99%
Chemical Name or Material 4'-Cyclohexylacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.