missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyclohexylacetophenone, 98%, ACROS Organics™

Product Code. 10566631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10566631 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10566631 Supplier Thermo Scientific Acros Supplier No. 339120050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 18594-05-3
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H18O
MDL Number MFCD00001453
Quantity 5 g
Synonym 4'-cyclohexylacetophenone, 1-4-cyclohexylphenyl ethanone, 1-4-cyclohexylphenyl ethan-1-one, 4-cyclohexylacetophenone, ethanone, 1-4-cyclohexylphenyl, 1-acetyl-4-cyclohexylbenzene, p-cyclohexylacetophenone, acmc-209ena, 4-cyclohexylace-tophenone, 4'-cyclohexyl actephenone
InChI Key MSDQNIRGPBARGC-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=C(C=C1)C1CCCCC1
IUPAC Name 1-(4-cyclohexylphenyl)ethan-1-one
Molecular Weight (g/mol) 202.30
PubChem CID 87715
Formula Weight 202.3
Chemical Name or Material 4'-Cyclohexylacetophenone
Show More Show Less
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.