missing translation for 'onlineSavingsMsg'
Learn More

(4-Cyclohexyl-phenyl)-(4-isopropyl-phenyl)-amine, TRC

Product Code. 30357917
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357917 500 mg -
30365551 50 mg 50mg
30350485 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357917 Supplier Toronto Research Chemicals Supplier No. TRCC996185500MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886365-92-0
Molecular Formula C21H27N
Molecular Weight (g/mol) 293.45
IUPAC Name 4-cyclohexyl-N-(4-propan-2-ylphenyl)aniline
SMILES CC(C)c1ccc(Nc2ccc(cc2)C3CCCCC3)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C21H27N/c1-16(2)17-8-12-20(13-9-17)22-21-14-10-19(11-15-21)18-6-4-3-5-7-18/h8-16,18,22H,3-7H2,1-2H3
Formula Weight 293.2143
Chemical Name or Material (4-Cyclohexyl-phenyl)-(4-isopropyl-phenyl)-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.