missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanopyridine, 98%, Thermo Scientific Chemicals

Catalog Number 11364739
Click to view available options
Quantity:
10 g
100 g
500 g
2500 g
This item is not returnable. View return policy
100-48-1
C6H4N2
104.112
MFCD00006417
GPHQHTOMRSGBNZ-UHFFFAOYSA-N
4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
7506
CHEBI:28020
pyridine-4-carbonitrile
C1=CN=CC=C1C#N
This item is not returnable. View return policy
100-48-1
C6H4N2
104.112
MFCD00006417
GPHQHTOMRSGBNZ-UHFFFAOYSA-N
4-cyanopyridine, isonicotinonitrile, 4-pyridinecarbonitrile, isonicotinic acid nitrile, 4-pyridinenitrile, 4-cyano pyridine, .gamma.-cyanopyridine, gamma-cyanopyridine, p-cyanopyridine, pubchem11155
7506
CHEBI:28020
pyridine-4-carbonitrile
C1=CN=CC=C1C#N

4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyanopyridine is used as an intermediate in organic synthesis and pharmaceutical substances like isonicotinylhydrazide, which is used in the treatment of tuberculosis. It is used as a precursor for the preparation of isonicotinic acid and 4-diemthylaminopyridine (DMAP). It is involved in the synthesis of 6-methyl-2-pyridin-4-yl-pyrimidin-4-ylamine by reacting with acetonitrile.

Solubility
Soluble in water.

Notes
Incompatible with strong acids, strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 76°C to 80°C
pH 6
Boiling Point 194°C to 196°C
Flash Point 88°C (190°F)
Odor Pungent
Quantity 10 g
Beilstein 107712
Solubility Information Soluble in water.
Formula Weight 104.11
Percent Purity 98%
Chemical Name or Material 4-Cyanopyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'