missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanopiperidine, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
4-Cyanopiperidine is used as an intermediate for the preparation of pharmaceutical substances like antidepressants and anti-inflammatory. It is used as a precursor for the synthesis of piperidine derivatives.
Solubility
Soluble in chloroform.
Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Density | 0.98 |
Boiling Point | 226°C |
Flash Point | 91°C (196°F) |
Odor | Amine-like |
Refractive Index | 1.4755 |
Quantity | 5 g |
UN Number | UN3276 |
Solubility Information | Soluble in chloroform. |
Formula Weight | 110.16 |
Percent Purity | 96% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'