Learn More
4-Cyanophenylacetic acid, 97%
Description
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Cyanophenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.
Solubility
Soluble in CHCl3, THF, dioxane.
Notes
Storage temperature is 2 - 4°C and Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
| Melting Point | 150°C to 152°C |
| Quantity | 1 g |
| UN Number | UN3439 |
| Solubility Information | Soluble in CHCl3,THF,dioxane. |
| Formula Weight | 161.16 |
| Percent Purity | 97% |
| Chemical Name or Material | 4-Cyanophenylacetic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.