missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanophenol-2,3,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxybenzonitrile-D4 |
| CAS | 1025089-21-7 |
| Molecular Formula | C7 D4 H N O |
| Quantity | 0.05 g |
| Synonym | Benzonitrile-2,3,5,6-d4, 4-hydroxy- (ACI), 4-Hydroxybenzonitrile-2,3,5,6-d4 (ACI), 4-Hydroxybenzonitrile-D4, 4-Cyanophenol-2,3,5,6-d4 |
| InChI Formula | InChI=1 S/C7H5NO/c8-5-6-1-3-7(9)4-2-6/h1-4,9 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C#N)c([2 H])c([2 H])c1O |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-hydroxybenzonitrile |
| Molecular Weight (g/mol) | 123.1454 |
| Formula Weight | 123.0622 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction