missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog Number 15497026
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1220219-61-3
C14H17BFNO2
261.103
SAFMDYCIEJEGJQ-UHFFFAOYSA-N
2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, acmc-209ajh, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile
54759097
2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC#N)F
This item is not returnable. View return policy
1220219-61-3
C14H17BFNO2
261.103
SAFMDYCIEJEGJQ-UHFFFAOYSA-N
2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, acmc-209ajh, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl acetonitrile, 2-2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl acetonitrile
54759097
2-[2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]acetonitrile
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)CC#N)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 261.1
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Cyanomethyl-3-fluorobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'