missing translation for 'onlineSavingsMsg'
Learn More

4-(Cyanomethoxy)phenylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. p-4380081 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11916141

Brand: Thermo Scientific Acros 445400010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 947533-23-5
Molecular Formula C8H8BNO3
Molecular Weight (g/mol) 176.97
MDL Number MFCD09908170
InChI Key GKJPNYHKHSLHCC-UHFFFAOYSA-N
Synonym 4-cyanomethoxyphenylboronic acid, 4-cyanomethoxy phenyl boronic acid, 4-cyanomethoxy phenylboronic acid, acmc-209rtj
PubChem CID 21918429
IUPAC Name [4-(cyanomethoxy)phenyl]boronic acid
SMILES OB(O)C1=CC=C(OCC#N)C=C1

Specifications

Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 176.97
Percent Purity 97%
Chemical Name or Material 4-(Cyanomethoxy)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(Cyanomethoxy)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.