missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzyl Bromide, TRC

Product Code. 30349317
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349317 Supplier Toronto Research Chemicals Supplier No. TRCC9623005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Cyanobenzyl Bromide
CAS 17201-43-3
Molecular Formula C8 H6 Br N
Quantity 5 g
Synonym Benzonitrile, 4-(bromomethyl)-, p-Tolunitrile, α-bromo- (7CI,8CI), 4-(Bromomethyl)benzonitrile, (4-Cyanophenyl)methyl bromide, (p-Cyanophenyl)bromomethane, 1-Bromomethyl-4-cyanobenzene, 4-Cyanobenzyl bromide, NSC 95792, p-(Bromomethyl)benzonitrile, p-Cyanobenzyl bromide, α-Bromo-p-toluenecarbonitrile, α-Bromo-p-tolunitrile, α-Bromo-p-tolylnitrile
InChI Formula InChI=1S/C8H6BrN/c9-5-7-1-3-8(6-10)4-2-7/h1-4H,5H2
SMILES BrCc1ccc(cc1)C#N
IUPAC Name 4-(bromomethyl)benzonitrile
Molecular Weight (g/mol) 196.04
Formula Weight 194.9684
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.