missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzophenone, 98%

Product Code. 11482418
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482418 1 g 1g
11492418 5 g 5g
11402428 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482418 Supplier Thermo Scientific Alfa Aesar Supplier No. A15764.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1503-49-7 | C14H9NO | 207.232 g/mol

Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)quadricyclane(Q) and QN isomerization. It is employed as a triplet sensitizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Substituted benzophenones are used to determine the quantum yields for norbornadiene(N)→quadricyclane(Q) and Q→N isomerization. It is employed as a triplet sensitizers.

Solubility
Insoluble in water.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1503-49-7
Molecular Formula C14H9NO
Molecular Weight (g/mol) 207.232
MDL Number MFCD00016382
InChI Key YSZWJJANSNFQMM-UHFFFAOYSA-N
PubChem CID 73921
IUPAC Name 4-benzoylbenzonitrile
SMILES C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)C#N

Specifications

Melting Point 110°C to 114°C
Quantity 1 g
UN Number UN3439
Beilstein 641181
Synonym 4-cyanobenzophenone, 4-benzoyl-benzonitrile, benzonitrile, 4-benzoyl, 4-cyano-benzophenon, 4-cyano-benzophenone, acmc-20amk8, #, benzonitrile,4-benzoyl, 4-phenylcarbonyl benzenecarbonitrile
Solubility Information Insoluble in water.
Formula Weight 207.23
Percent Purity 98%
Chemical Name or Material 4-Cyanobenzophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.