missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzenesulfonyl chloride, 97%

Product Code. 11498016
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498016 25 g 25g
11478016 1 g 1g
11488016 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498016 Supplier Thermo Scientific Alfa Aesar Supplier No. A14081.14
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 49584-26-1 | C7H4ClNO2S | 201.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49584-26-1
Molecular Formula C7H4ClNO2S
Molecular Weight (g/mol) 201.62
MDL Number MFCD00052337
InChI Key DBMFYTQPPBBKHI-UHFFFAOYSA-N
Synonym 4-cyanobenzene-1-sulfonyl chloride, 4-cyanobenzenesulfonylchloride, p-cyanobenzenesulphonyl chloride, 4-cyano benzenesulfonyl chloride, benzenesulfonyl chloride, 4-cyano, 4-cyanophenylsulfonyl chloride, p-cyanobenzenesulfonyl chloride, 4-cyanobenzenesulphonyl chloride, 4-cyano-benzenesulfonyl chloride, 4-chlorosulfonyl benzenecarbonitrile
PubChem CID 39497
SMILES ClS(=O)(=O)C1=CC=C(C=C1)C#N

Specifications

Melting Point 107°C to 110°C
Quantity 25 g
UN Number UN3261
Beilstein 777939
Sensitivity Moisture sensitive
IUPAC Name 4-cyanobenzenesulfonyl chloride
Formula Weight 201.63
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.