missing translation for 'onlineSavingsMsg'
Learn More

4-Cyanobenzeneboronic acid pinacol ester, 97%

Product Code. 11367629
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367629 1 g 1g
11377629 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367629 Supplier Thermo Scientific Alfa Aesar Supplier No. H28208.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 171364-82-2 | C13H16BNO2 | 229.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171364-82-2
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.09
MDL Number MFCD03093897
InChI Key HOPDTPGXBZCBNP-UHFFFAOYSA-N
Synonym 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyanophenylboronic acid pinacol ester, 4-cyanobenzeneboronic acid pinacol ester, 4-cyanophenylboronic acid, pinacol ester, benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-cyanobenzeneboronic acid, pinacol ester, 2-4-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 4-cyanophenylboronicacidpinacolester
PubChem CID 2734625
IUPAC Name 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C#N

Specifications

Quantity 1 g
Formula Weight 229.08
Percent Purity 97%
Chemical Name or Material 4-Cyanobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.