missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyanoacetanilide, 98%, Thermo Scientific™

Catalog Number p-3212054
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
35704-19-9
C9H8N2O
160.18
MFCD00001814
UFKRTEWFEYWIHD-UHFFFAOYSA-N
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
37256
N-(4-cyanophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)C#N
This item is not returnable. View return policy
35704-19-9
C9H8N2O
160.18
MFCD00001814
UFKRTEWFEYWIHD-UHFFFAOYSA-N
n-4-cyanophenyl acetamide, 4-cyanoacetanilide, 4-acetamidobenzonitrile, acetamide, n-4-cyanophenyl, 4'-cyanoacetanilide, p-acetamidobenzonitirle, acetanilide, 4'-cyano, p-acetylamino-benzonitrile, p-cyanoacetanilide, p-acetaminobenzonitrile
37256
N-(4-cyanophenyl)acetamide
CC(=O)NC1=CC=C(C=C1)C#N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 204°C to 208°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CONHC6H4CN
Quantity 5 g
Formula Weight 160.18
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 4'-Cyanoacetanilide
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Cyanoacetanilide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'