missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-L-phenylalanine, 95%

Product Code. 15460329
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460329 250 mg 250mg
15440329 1 g 1g
15450329 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460329 Supplier Thermo Scientific Alfa Aesar Supplier No. H63572.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 167479-78-9 | C10H10N2O2 | 190.202 g/mol

4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-L-phenylalanine used as a intermediate in organic synthesis, pharmaceutical, chemical research and biochemical application.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167479-78-9
Molecular Formula C10H10N2O2
Molecular Weight (g/mol) 190.202
MDL Number MFCD00270363
InChI Key KWIPUXXIFQQMKN-VIFPVBQESA-N
Synonym l-4-cyanophenylalanine, 4-cyano-l-phenylalanine, s-2-amino-3-4-cyanophenyl propanoic acid, h-phe 4-cn-oh, 2s-2-amino-3-4-cyanophenyl propanoic acid, 4-cyanophenylalanine, s-2-amino-3-4-cyanophenyl propionic acid, d-4-cn-phe-oh
PubChem CID 1501865
IUPAC Name (2S)-2-amino-3-(4-cyanophenyl)propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)C#N

Specifications

UN Number UN3439
Solubility Information Slightly soluble in water.
Quantity 250 mg
Formula Weight 190.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Cyano-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.