missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4-phenylpiperidine hydrochloride, 97%

Product Code. 11462040
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462040 10 g 10g
11452040 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11462040 Supplier Thermo Scientific Alfa Aesar Supplier No. H28414.09
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51304-58-6 | C12H15ClN2 | 222.716 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51304-58-6
Molecular Formula C12H15ClN2
Molecular Weight (g/mol) 222.716
MDL Number MFCD00012775
InChI Key CQPHZBOPSZGTJM-UHFFFAOYSA-N
Synonym 4-cyano-4-phenylpiperidine hydrochloride, 4-phenylpiperidine-4-carbonitrile hydrochloride, 4-cyano-4-phenylpiperidinehydrochloride, 4-phenylpiperidine-4-carbonitrile, chloride, 4-phenylpiperidine-4-carbonitrile hcl, pubchem19077, acmc-20a3zw, 4-cyano-4-phenylpiperidine hcl, timtec-bb sbb003334, 4-phenylisonipecotonitrile hydrochloride
PubChem CID 3084973
IUPAC Name 4-phenylpiperidine-4-carbonitrile;hydrochloride
SMILES C1CNCCC1(C#N)C2=CC=CC=C2.Cl

Specifications

Melting Point 210°C to 214°C
Quantity 10 g
UN Number UN3439
Formula Weight 222.71
Percent Purity 97%
Chemical Name or Material 4-Cyano-4-phenylpiperidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.