missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-4-phenylcyclohexanone, 97%

Product Code. 15498829
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498829 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498829 Supplier Thermo Scientific Alfa Aesar Supplier No. L05281.06
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 25115-74-6 | C13H13NO | 199.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 25115-74-6
Melting Point 112°C to 118°C
Molecular Formula C13H13NO
MDL Number MFCD00044816
Quantity 5 g
Beilstein 2645593
Synonym 4-cyano-4-phenylcyclohexanone, 4-oxo-1-phenylcyclohexanecarbonitrile, 4-oxo-1-phenylcyclohexanenitrile, cyclohexanecarbonitrile, 4-oxo-1-phenyl, 4-oxo-1-phenyl-cyclohexanecarbonitrile, 4-oxo-1-phenyl-1-cyclohexanecarbonitrile, acmc-1ciss, 4-cyano-4phenylcyclohexanone, 4-phenyl 4-cyanocylohexanone, 4-phenyl 4-cyanocyclohexanone
InChI Key GKXOABVSZWCJJK-UHFFFAOYSA-N
SMILES O=C1CCC(CC1)(C#N)C1=CC=CC=C1
IUPAC Name 4-oxo-1-phenylcyclohexane-1-carbonitrile
Molecular Weight (g/mol) 199.25
PubChem CID 91282
Formula Weight 199.24
Percent Purity 98%
Chemical Name or Material 4-Cyano-4-phenylcyclohexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.