missing translation for 'onlineSavingsMsg'
Learn More

4'-Cyano-4-biphenylyl trans-4-n-propylcyclohexanecarboxylate, 99%, Thermo Scientific™

Product Code. 15402726
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67284-57-5
Molecular Formula C23H25NO2
Molecular Weight (g/mol) 347.458
MDL Number MFCD01076321
InChI Key YVEDSNAPDAXLDD-UHFFFAOYSA-N
Synonym 4'-cyanobiphenyl-4-yl trans-4-propylcyclohexanecarboxylate, 4-4-cyanophenyl phenyl 4-propylcyclohexane-1-carboxylate, trans-4-propyl-cyclohexanecarboxylic acid 4'-cyano-biphenyl-4-yl ester, 4'-cyano-4-biphenylyl trans-4-n-propylcyclohexanecarboxylate, 4-propylcyclohexanecarboxylic acid 4'-cyanobiphenyl-4-yl ester, 4'-cyano 1,1'-biphenyl-4-yl 4-propylcyclohexane-1-carboxylate, trans-4'-cyano-1,1'-biphenyl-4-yl 4-propylcyclohexanecarboxylate, 4-propyl-1-cyclohexanecarboxylic acid 4-4-cyanophenyl phenyl ester
PubChem CID 15044673
IUPAC Name [4-(4-cyanophenyl)phenyl] 4-propylcyclohexane-1-carboxylate
SMILES CCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)C3=CC=C(C=C3)C#N

Specifications

Quantity 1 g
Formula Weight 347.46
Percent Purity 99%
Chemical Name or Material 4'-Cyano-4-biphenylyl trans-4-n-propylcyclohexanecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'