missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3-fluoropyridine, 95%

Product Code. 15478148
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478148 5 g 5g
15468148 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15478148 Supplier Thermo Scientific Alfa Aesar Supplier No. H61594.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 113770-88-0 | C6H3FN2 | 122.102 g/mol

4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-3-fluoropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113770-88-0
Molecular Formula C6H3FN2
Molecular Weight (g/mol) 122.102
MDL Number MFCD04112524
InChI Key RFQHDTRSEWCXJP-UHFFFAOYSA-N
PubChem CID 22156440
IUPAC Name 3-fluoropyridine-4-carbonitrile
SMILES C1=CN=CC(=C1C#N)F

Specifications

Color White to Yellow
Quantity 5 g
UN Number UN3439
Synonym 3-fluoro-4-cyanopyridine, 3-fluoroisonicotinonitrile, 3-fluoro-4-cyano-pyridine, 3-fluoro-isonicotinonitrile, 4-pyridinecarbonitrile,3-fluoro, 4-pyridinecarbonitrile, 3-fluoro, 4-pyridinecarbonitrile,3-fluoro-9ci, 3-fluoro-4-pyridinecarbonitrile, 3-fluoranylpyridine-4-carbonitrile, 4-pyridinecarbonitrile, 3-fluoro-9ci
Solubility Information Slightly soluble in water.
Formula Weight 122.1
Percent Purity 95%
Physical Form Crystalline
Chemical Name or Material 4-Cyano-3-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.