Learn More
4-Cyano-3-fluorophenylboronic acid, 97%, Thermo Scientific Chemicals
£38.30 - £370.17
Chemical Identifiers
CAS | 843663-18-3 |
---|---|
Molecular Formula | C7H5BFNO2 |
Molecular Weight (g/mol) | 164.93 |
MDL Number | MFCD03411549 |
InChI Key | DECWLXUOZUMPBF-UHFFFAOYSA-N |
Synonym | 4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug |
PubChem CID | 2783130 |
IUPAC Name | (4-cyano-3-fluorophenyl)boronic acid |
SMILES | OB(O)C1=CC(F)=C(C=C1)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13488608
|
Thermo Scientific Acros
448710010 |
1 g |
£38.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13498608
|
Thermo Scientific Acros
448710050 |
5 g |
£117.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13408618
|
Thermo Scientific Acros
448710250 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
843663-18-3 | |
164.93 | |
DECWLXUOZUMPBF-UHFFFAOYSA-N | |
2783130 | |
OB(O)C1=CC(F)=C(C=C1)C#N |
C7H5BFNO2 | |
MFCD03411549 | |
4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug | |
(4-cyano-3-fluorophenyl)boronic acid |
Specifications
843663-18-3 | |
97% | |
C7H5BFNO2 | |
4-cyano-3-fluorobenzeneboronic acid, 4-cyano-3-fluorophenyl boronic acid, 4-borono-2-fluorobenzonitrile, 4-cyano-3-fluoro-phenyl boronic acid, 4-cyano-3-fluoro phenyl boronic acid, 3-fluoro-4-cyanophenylboronic acid, boronic acid, 4-cyano-3-fluorophenyl, 4-cyano-3-fluorophenylboronicacid, pubchem1801, acmc-209pug | |
OB(O)C1=CC(F)=C(C=C1)C#N | |
164.93 | |
164.93 | |
4-Cyano-3-fluorophenylboronic acid |
362°C to 366°C | |
Glass bottle | |
MFCD03411549 | |
DECWLXUOZUMPBF-UHFFFAOYSA-N | |
(4-cyano-3-fluorophenyl)boronic acid | |
2783130 | |
97% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e
GHS Signal Word: Warning
- 4-Cyano-3-fluorophenylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only