missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Product Code. 15407886
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003298-73-4
Molecular Formula C13H14BF2NO2
Molecular Weight (g/mol) 265.07
MDL Number MFCD22419275
InChI Key XYMVJTWEERDWFY-UHFFFAOYSA-N
Synonym 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 4-cyano-3,5-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, 2,6-difluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzonitrile
PubChem CID 58504590
IUPAC Name 2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=C(C#N)C(F)=C1

Specifications

Quantity 250 mg
Formula Weight 265.06
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 4-Cyano-3,5-difluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'