missing translation for 'onlineSavingsMsg'
Learn More

4-Cyano-2-fluorobenzoic acid, 98%

Product Code. 11395555
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11395555

Brand: Thermo Scientific Alfa Aesar L19722.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 164149-28-4 | C8H4FNO2 | 165.12 g/mol

4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Cyano-2-fluorobenzoic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 164149-28-4
Molecular Formula C8H4FNO2
Molecular Weight (g/mol) 165.12
MDL Number MFCD03094454
InChI Key KEJMSTJTAWACNI-UHFFFAOYSA-N
Synonym 2-fluoro-4-cyanobenzoic acid, benzoic acid, 4-cyano-2-fluoro, 4-cyano-2-fluoro-benzoic acid, 4-cyano-2-fluorobenzoicacid, pubchem4603, acmc-1bzbk, intermediates-zcf02232, 2-fluoro4-cyanobenzoic acid, ksc489o1d, 4-carboxy-3-fluorobenzonitrile
PubChem CID 590659
IUPAC Name 4-cyano-2-fluorobenzoic acid
SMILES OC(=O)C1=C(F)C=C(C=C1)C#N

Specifications

Melting Point 214°C to 218°C
Quantity 5 g
Beilstein 8051929
Solubility Information Insoluble in water.
Formula Weight 165.12
Percent Purity 98%
Chemical Name or Material 4-Cyano-2-fluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.