missing translation for 'onlineSavingsMsg'
Learn More

4-Cumylphenol, TRC

Product Code. 30010449
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010449 500 mg 500mg
30004538 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010449 Supplier Toronto Research Chemicals Supplier No. TRCC834580500MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 599-64-4
Molecular Formula C15 H16 O
Molecular Weight (g/mol) 212.29
Synonym p-(α,α-Dimethylbenzyl)-phenol, 4-(1-Methyl-1-phenylethyl)phenol, 2-(4-Hydroxyphenyl)-2-phenylpropane, 2-Phenyl-2-(4-hydroxyphenyl)propane, 4-(Dimethylphenylmethyl)phenol, 4-(α,α-Dimethylbenzyl)phenol, 4-Hydroxydiphenyldimethylmethane, 4-α-Cumylphenol, NSC 6237, PCP, p-(α,α-Dimethylbenzyl)phenol, p-(α-Cumyl)phenol, p-Cumylphenol
IUPAC Name 4-(1-methyl-1-phenyl-ethyl)phenol
SMILES CC(C)(c1ccccc1)c2ccc(O)cc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C15H16O/c1-15(2,12-6-4-3-5-7-12)13-8-10-14(16)11-9-13/h3-11,16H,1-2H3
Formula Weight 212.1201
Percent Purity >95%
Chemical Name or Material 4-Cumylphenol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.