missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanone, 98+%

Product Code. 11413584
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413584 5 g 5g
11423584 25 g 25g
11433584 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413584 Supplier Thermo Scientific Alfa Aesar Supplier No. A11242.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 491-37-2 | C9H8O2 | 148.161 g/mol

4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chromanone has been used to study the substrate specificity and catalytic ability of 4-hydroxyacetophenone monooxygenase isolated from Pseudomonas fluorescens ACB

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents and moisture. Store away from oxidizing agents. Store at 2-8°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-37-2
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.161
MDL Number MFCD00006840
InChI Key MSTDXOZUKAQDRL-UHFFFAOYSA-N
Synonym 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone
PubChem CID 68110
IUPAC Name 2,3-dihydrochromen-4-one
SMILES C1COC2=CC=CC=C2C1=O

Specifications

Melting Point 36°C to 40°C
Boiling Point 127°C to 128°C (13 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.575
Quantity 5 g
Beilstein 124652
Solubility Information Slightly soluble in water.
Formula Weight 148.16
Percent Purity ≥98%
Chemical Name or Material 4-Chromanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.