missing translation for 'onlineSavingsMsg'
Learn More

4-Chromanone, 96%

Product Code. 10347280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347280 50 g Glass bottle 50g
10295860 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347280 Supplier Thermo Scientific Acros Supplier No. 153460500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 491-37-2 | C9H8O2 | 148.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-37-2
Molecular Formula C9H8O2
Molecular Weight (g/mol) 148.16
MDL Number MFCD00006840
InChI Key MSTDXOZUKAQDRL-UHFFFAOYSA-N
Synonym 4-chromanone, chroman-4-one, chromanone, 4h-1-benzopyran-4-one, 2,3-dihydro, 2,3-dihydro-4h-chromen-4-one, 2,3-dihydro-1-benzopyran-4-one, isochromanone, 3,4-dihydro-2h-1-benzopyran-4-one, 2,3-dihydro-4h-1-benzopyran-4-one, dihydrochromone
PubChem CID 68110
IUPAC Name 2,3-dihydrochromen-4-one
SMILES C1COC2=CC=CC=C2C1=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 34°C to 40°C
Color White to Yellow
Boiling Point 127°C to 128°C (13.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.575
Quantity 50 g
Beilstein 17, II, 335
Formula Weight 148.16
Percent Purity 96%
Physical Form Low Melting Mass, Crystals or Crystalline Powder
Chemical Name or Material 4-Chromanone, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.