missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiophenol, 97%

Product Code. 11410318
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410318 25 g 25g
11420318 100 g 100g
11430318 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410318 Supplier Thermo Scientific Alfa Aesar Supplier No. B23838.14
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 106-54-7 | C6H5ClS | 144.616 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-54-7
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.616
MDL Number MFCD00004847
InChI Key VZXOZSQDJJNBRC-UHFFFAOYSA-N
Synonym 4-chlorothiophenol, p-chlorothiophenol, benzenethiol, 4-chloro, p-chlorobenzenethiol, p-chlorthiofenol, p-chlorophenylmercaptan, benzenethiol, p-chloro, p-mercaptochlorobenzene, 4-chlorophenylmercaptan, 1-chloro-4-mercaptobenzene
PubChem CID 7815
IUPAC Name 4-chlorobenzenethiol
SMILES C1=CC(=CC=C1S)Cl

Specifications

Melting Point 48°C to 52°C
Boiling Point 205°C to 207°C
Flash Point 81°C (177°F)
Quantity 25 g
UN Number UN1759
Beilstein 605971
Sensitivity Air Sensitive
Formula Weight 144.62
Percent Purity 97%
Chemical Name or Material 4-Chlorothiophenol
compliance-icons compliance-icons
Product Identifier
  • 4-Chlorothiophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: p-Chlorothiophenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.