missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiobenzamide, 97%

Product Code. 11384739
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
Unit Size:
100g
10g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384739 10 g 10g
11409882 25 g 25g
11394739 50 g 50g
11419882 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11384739 Supplier Thermo Scientific Alfa Aesar Supplier No. A12021.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2521-24-6 | C7H6ClNS | 171.64 g/mol

4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2521-24-6
Molecular Formula C7H6ClNS
Molecular Weight (g/mol) 171.64
MDL Number MFCD00040956
InChI Key OKPUICCJRDBRJT-UHFFFAOYSA-N
Synonym 4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
PubChem CID 2734826
SMILES NC(=S)C1=CC=C(Cl)C=C1

Specifications

Melting Point 123°C to 129°C
Quantity 10 g
Beilstein 606603
Solubility Information Insoluble in water.
IUPAC Name 4-chlorobenzenecarbothioamide
Formula Weight 171.65
Percent Purity 97%
Chemical Name or Material 4-Chlorothiobenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.