missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[3,2-d]pyrimidine, 97%, Thermo Scientific™

Product Code. 10367064
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10367064 25 g Amber Glass Bottle 25g
10533032 1 g Amber Glass Bottle 1g
10487612 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367064 Supplier Thermo Scientific Maybridge Supplier No. GK00377EB
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 16269-66-2
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.614
MDL Number MFCD00122144
InChI Key TWTODSLDHCDLDR-UHFFFAOYSA-N
Synonym 4-chlorothieno 3,2-d pyrimidine, thieno 3,2-d pyrimidine, 4-chloro, 4-chloro-thieno 3,2-d pyrimidine, 4-chlorothieno 3,2-d-pyrimidine, 4-chlorothiopheno 3,2-d pyrimidine, pubchem14662, 4-chlorothienopyrimidine, acmc-209dov
PubChem CID 2735821
IUPAC Name 4-chlorothieno[3,2-d]pyrimidine
SMILES C1=CSC2=C1N=CN=C2Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 170.62
Percent Purity 97%
Chemical Name or Material 4-Chlorothieno[3, 2-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.