missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothieno[2,3-d]pyrimidine, Tech., Thermo Scientific™

Product Code. 10351735
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10351735 5 g 5g
10064662 1 g 1g
10046252 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10351735 Supplier Thermo Scientific Maybridge Supplier No. GK02802DE
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14080-59-2
Molecular Formula C6H3ClN2S
Molecular Weight (g/mol) 170.61
MDL Number MFCD01312373
InChI Key NZCRUBBNZGVREM-UHFFFAOYSA-N
PubChem CID 736618
IUPAC Name 4-chlorothieno[2,3-d]pyrimidine
SMILES ClC1=C2C=CSC2=NC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 4-chlorothieno 2,3-d pyrimidine, thieno 2,3-d pyrimidine, 4-chloro, 4-chloro-thieno 2,3-d pyrimidine, 4-chlorothiopheno 2,3-d pyrimidine, pubchem14686, acmc-1bujx, ksc522g5h, 4-chloro-thieno 2,3-d-pyrimidine, thieno 2,3-d pyrimidine,4-chloro
Formula Weight 170.62
Percent Purity 97%
Grade Technical
Chemical Name or Material 4-Chlorothieno[2, 3-d]pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.