missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chlorostyrene, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorostyrene |
| CAS | 1073-67-2 |
| Molecular Formula | C8 H7 Cl |
| Quantity | 50 g |
| Synonym | Benzene, 1-chloro-4-ethenyl- (9CI, ACI), 1-Chloro-4-ethenylbenzene (ACI), Styrene, p-chloro- (8CI), (4-Chlorophenyl)ethene, 1-Chloro-4-vinylbenzene, 4-Chloro-1-ethenylbenzene, 4-Chlorostyrene, NSC 18603, p-Chlorostyrene, p-Chlorovinylbenzene, p-Vinylphenyl chloride |
| InChI Formula | InChI=1S/C8H7Cl/c1-2-7-3-5-8(9)6-4-7/h2-6H,1H2 |
| SMILES | Clc1ccc(C=C)cc1 |
| IUPAC Name | 1-chloro-4-vinyl-benzene |
| Molecular Weight (g/mol) | 138.594 |
| Formula Weight | 138.0236 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction