missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorostyrene, 98+%, stab

Product Code. 11418877
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418877 10 g 10g
11428877 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418877 Supplier Thermo Scientific Alfa Aesar Supplier No. B20512.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073-67-2 | C8H7Cl | 138.59 g/mol

4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

Solubility
Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents, halogens, metal salts and polymerizing initiators. Store in a cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073-67-2
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.59
MDL Number MFCD00000632
InChI Key KTZVZZJJVJQZHV-UHFFFAOYSA-N
Synonym 4-chlorostyrene, p-chlorostyrene, 1-chloro-4-vinylbenzene, parachlorostyrene, benzene, 1-chloro-4-ethenyl, styrene, p-chloro, styrene, 4-chloro, unii-t0j05u220f, para-chlorostyrene
PubChem CID 14085
IUPAC Name 1-chloro-4-ethenylbenzene
SMILES ClC1=CC=C(C=C)C=C1

Specifications

Melting Point -16°C
Density 1.089
Boiling Point 191°C to 192°C
Flash Point 67°C (153°F)
Odor Characteristic
Refractive Index 1.565
Quantity 10 g
Beilstein 1098859
Solubility Information Immiscible with water.
Formula Weight 138.6
Percent Purity 99%
Chemical Name or Material 4-Chlorostyrene, stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.